Masterflex I/P® Variable-Speed Hazardous-Duty Drive, 10 to 430 rpm; 115V

Representative image only.
Masterflex Item # EW-07583-50
Pump safely in hazardous
environments
  • Use either L/S or I/P pump head on drive
  • L/S flow rate 0.6 to 2500 mL/min or I/P flow rate 60 to 8800 mL/min— flow rate depends on drive rpm, pump head, and tubing size
  • Fully mechanical speed control—adjust with lever then lock setting
  • UL-listed for Class 1, Groups C and D, Division 1 hazardous locations

$4,010.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Pump SeriesI/P
  • Control TypeAnalog Variable Speed
  • Min RPM (rpm)10
  • Max RPM (rpm)430
  • SpeedVariable
  • Speed Control±10%
  • Max Flow Rate (LPM)11.5
  • Min Flow Rate (LPM)0.06
  • Max Flow Rate (mL/min)11500
  • Min Flow Rate (mL/min)60
  • Reversible MotorNo
  • No. of Pump Heads Accepted2
  • Type of Pump Heads AcceptedI/P: Standard, Easy-Load, High-Performance; L/S: Standard, Quick Load, Easy-Load, Easy-Load II, High-Performance, Cartridge
  • Motor (watts)190
  • Drive IP ratingIP 21
  • Power (VAC)115
  • Power (amps)4.7
  • Power (Hz)60
  • Drive Length (in)22.0625
  • Drive Width (in)13.625
  • Drive Height (in)8.75
  • Drive Length (mm)560
  • Drive Width (mm)346
  • Drive Height (mm)223
  • Open-Head SensorNo
  • Housing MaterialPainted Steel
  • Motor hp1/3
  • DescriptionI/P variable-speed hazardous-duty drive, 10-430 rpm, 115V.
Two year warranty

Key Features

  • Use either L/S or I/P pump head on drive
  • L/S flow rate 0.6 to 2500 mL/min or I/P flow rate 60 to 8800 mL/min— flow rate depends on drive rpm, pump head, and tubing size
  • Fully mechanical speed control—adjust with lever then lock setting
  • UL-listed for Class 1, Groups C and D, Division 1 hazardous locations

More About this Item

Choose one of these variable-speed hazardous-duty drives for hazardous locations where special electrical precautions are necessary. Power switch and line cord are not included. Safely connect power using your own protected connections.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat