Masterflex Inert in-line inlet check valve; 0.014 Cv, PEEK/Chemraz
Representative image only.
Masterflex Item # EW-01355-16
No maintenance required!
  • Ideal for high-purity, low pressure
  • No metal components — inert flow path
  • No back flow leakage with back pressure of 0 to 100 psi
  • 1/4-28 UNF threaded connections
PRODUCT OPTIONS
$172.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/4"-28 UNF
  • Connection TypeUNF
  • Inlet Port1/4"-28 UNF
  • Breaking Pressure (PSI)0.23
  • Cv0.014
  • Max Pressure (PSI)100
  • Wetted MaterialsPEEK
  • DescriptionInert in-line inlet check valve; 0.014 Cv, PEEK/Chemraz
Key Features
  • Ideal for high-purity, low pressure
  • No metal components — inert flow path
  • No back flow leakage with back pressure of 0 to 100 psi
  • 1/4-28 UNF threaded connections
More About this Item
Unlike spring-actuated check valves that may restrict or impede the flow path, this in-line check valve features a smooth flow path that minimizes shear and turbulence.