Masterflex L/S PTFE-Diaphragm Pump System; 6 to 600 rpm, 90 to 260 VAC

Representative image only.
Masterflex Item # EW-77915-10
Ideal for chemical feed/metering and applications requiring high-purity, high-pressure or both
  • Flow range: 80 to 800 mL/min
  • Pump at pressures up to 50 psi (3.4 bar)
  • PTFE wetted parts help maintain fluid purity
  • Pump through filters or into pressurized system
  • Check valve on inlet side of pump holds prime for up to two hours when pump is off

$3,040.00USD / Each

$170.00USD / Each

for an additional
$170.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Pump SeriesL/S
  • Control TypeVariable-Speed Digital
  • Min RPM6
  • Max RPM600
  • Number of Channels1
  • Max Flow Rate Per Channel (mL/min)800
  • Min Flow Rate Per Channel (mL/min)80
  • Drive07528-10 (L/S® Precision Variable-Speed Drive)
  • Pump Head07090-42 (L/S® PTFE-Diaphragm Pump Head)
  • Tubing06605-15, 12-ft (PTFE Tubing, 1/4" x 3/8")
  • Drive IP ratingIP 33
  • Power (VAC)90 to 260
  • Power (amps)1.8A at 115v; 1.2A at 230v
  • Power (Hz)50/60
  • Width (in)8.3
  • Height (in)8.5
  • Length (in)17.3
  • Width (cm)21.08
  • Height (cm)21.59
  • Length (cm)43.94
  • DescriptionL/S PTFE-Diaphragm Pump System; 6 to 600 rpm, 90 to 260 VAC

Includes

PTFE-diaphragm pump head 07090-42, 12 ft (3.6-m) of PTFE tubing (1/4" ID x 3/8" OD), Masterflex; L/S; precision variable-speed drive 07528-10 (6 to 600 rpm), two PFA pipe adapters (1/4" NPT(F) x 3/8" OD), foot valve, mounting tools, and coupling hardware.

Key Features

  • Flow range: 80 to 800 mL/min
  • Pump at pressures up to 50 psi (3.4 bar)
  • PTFE wetted parts help maintain fluid purity
  • Pump through filters or into pressurized system
  • Check valve on inlet side of pump holds prime for up to two hours when pump is off
  • Unique geometry minimizes fluid churning and dead volume for high metering accuracy without reagent contamination

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-847-549-7600
Live chat