Masterflex L/S® RS-232 Cable for L/S® Computer-Compatible Drives to PC; 8 ft
Representative image only.
Masterflex Item # EW-07550-64
Easily connect your Masterflex® L/S® computer-compatible drive to a PC
  • Featuring RS-232 connections with 8 foot cable length
$135.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Operating PlatformPC
  • DescriptionRS-232 interface cable; connects L/S computer-compatible drives to PC, 8 ft.
Key Features
  • Featuring RS-232 connections with 8 foot cable length
More About this Item
Monitor and control your Servodyne mixer system from an IBM® or 100% compatible computer via a computer interface. Software programs let you to control up to 16 Servodyne mixers and Masterflex® computer-compatible pumps. Various application programs let you dedicate your automated setup for mixing and diluting, sequence control, flow proportioning, reversible batch dispensing, delivering single and multiple slope gradients, or customizing procedures for specialized applications.