Masterflex Liquid Flow Switch for Threaded Plastic Piping, Open; 0.5 GPM
Representative image only.
Masterflex Item # EW-32774-62
Connects quickly to threaded plastic piping
  • Chemically resistant — wetted materials stainless steel and polypropylene
  • Effortlessly install with 1/2" NPT(M) connections
  • NSF and FDA approved
PRODUCT OPTIONS
$187.00USD / Each
$332.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure (PSI)100
  • Max Flow Rate (GPM)0.5
  • Wetted Materialspolypropylene, Viton®, 316 SS, and PH 15-7 Mo stainless steel
  • Max Operating Temperature (° F)212
  • Max Operating Temperature (° C)100
  • Process Connection (NPT(F))3/4
  • DescriptionLiquid flow switch for threaded plastic piping; 0.5 GPM
Key Features
  • Chemically resistant — wetted materials stainless steel and polypropylene
  • Effortlessly install with 1/2" NPT(M) connections
  • NSF and FDA approved
More About this Item
These normally open switches have 3/4" NPT(F) ports for a quick connection to threaded plastic piping. Polypropylene and stainless steel wetted parts offer excellent chemical resistance. The materials are also NSF approved and FDA compliant; approved for potable water treatment applications such as chlorinators, purifiers, and heaters. Switches include 24" lead wires.