Masterflex Mechanically Coupled Centrifugal Pump Flange Kit for 1 1/2" and 2" Connections
Representative image only.
Masterflex Item # EW-70725-72
Enhance the life of your pump with service flange kits
  • Fits 1 1/2" and 2" connections
PRODUCT OPTIONS
$469.10USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Wetted MaterialsSS flanges, gaskets, nuts, bolts, flat washer, lockwashers
  • DescriptionFlange Kit for 1-1/2" and 2" Connections
Includes
Stainless steel 150# threaded RF companion flanges, gaskets, nuts, bolts, flat washer, and lock washers.
Key Features
  • Fits 1 1/2" and 2" connections
More About this Item
Flange kits are competible with your 304 SS mechanically coupled centrifugal pumps.