Masterflex Open-Head Sensor Retrofit System Controller
Representative image only.
Masterflex Item # EW-77097-00
Instantly shuts down the drive if the pump head is opened during operation
  • Use in conjunction with Masterflex® L/S® and I/P® open-head sensor equipped pump heads
  • Compatible with Masterflex® L/S® and I/P® pumps without integrated open-head sensor
  • Controller accepts input signals from up to two pump heads
  • Protect technicians and operators tasked with tubing changes and routine maintenance in busy production environments
  • Controller is NEMA 4X / IP66 rated for washdown
$668.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • No. of Pump Heads Accepted2
  • Type of Pump Heads AcceptedUse only with I/P Easy-Load pump head 77602-30 and L/S Easy-Load II pump heads 77200-30 and 77200-32
  • Drive RequirementsCompatible with all I/P drives and L/S drives that accept the Easy-Load II pump head
  • Controller IP ratingIP 66
  • DescriptionOpen-Head Sensor Retrofit System Controller
Key Features
  • Use in conjunction with Masterflex® L/S® and I/P® open-head sensor equipped pump heads
  • Compatible with Masterflex® L/S® and I/P® pumps without integrated open-head sensor
  • Controller accepts input signals from up to two pump heads
  • Protect technicians and operators tasked with tubing changes and routine maintenance in busy production environments
  • Controller is NEMA 4X / IP66 rated for washdown
More About this Item
The retrofit system is compatible with all I/P® pumps and with L/S® pumps that accept the Easy-Load® II pump head.
Use only with I/P® Easy-Load® pump head 77602-30 and with L/S® Easy-Load® II pump heads 77200-30 and 77200-32.