Masterflex Optional/Replacement PTFE O-rings; 6/Pk
Representative image only.
Masterflex Item # EW-03235-20
Gilmont® syringe- and burette-style dispenser accessory
  • Make the most of your micrometer syringe and burette dispensers with these replacement parts and accessories
$34.00USD / Pkg of 6
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPTFE
  • Compatible With07840-00 (Gilmont GS-1100)
  • DescriptionOptional/Replacement PTFE O-rings; 6/Pk
Key Features
  • Make the most of your micrometer syringe and burette dispensers with these replacement parts and accessories
More About this Item
Ideal for use with Gilmont GS-1100 micrometer syringe.
For use with 07840-00 syringe-style dispenser.