Masterflex Paddle Wheel Micro-Flo Rate and Total Flowmeter, 3/8" OD; 3000 mL/min
Representative image only.
Masterflex Item # EW-32550-19
Economical low-flow ranges
  • Paddle wheel technology is accurate, affordable, and relatively easy to install and maintain
  • Ideally suited to open channels and partially filled pipes containing water
  • Displays flow rate and total
  • Preset factory calibrations or custom field-calibrated
PRODUCT OPTIONS
$437.90USD / Each
$332.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy6% full-scale
  • Max Pressure (PSI)200
  • Min Flow Rate (mL/min)300
  • Max Flow Rate (mL/min)3000
  • Material FittingPVDF
  • Material O-RingViton®
  • Material HousingPVDF
  • Max Operating Temperature (° F)130
  • Max Operating Temperature (° C)54
  • Process Connection1/4" ID x 3/8" OD tubing connection
  • Power115 VAC
  • DescriptionMicro-Flo Rate and Total Meter, 300-3000 mL/min, 3/8" OD
Key Features
  • Paddle wheel technology is accurate, affordable, and relatively easy to install and maintain
  • Ideally suited to open channels and partially filled pipes containing water
  • Displays flow rate and total
  • Preset factory calibrations or custom field-calibrated
More About this Item
These economical flowmeters are capable of very low flow rates. The display is programmable for preselected factory settings or can be field calibrated for high accuracy in a specific application. Displays units in milliliters, ounces, liters, or gallons; displays time in minutes, hours, or days. Display can be programmed for up to four decimal places. All units are supplied with a 115 VAC/DC power supply.