Masterflex Power Cord, 230 VAC, Australian Plug; 6-ft Long
Representative image only.
Masterflex Item # EW-50001-60
Optimize performance of your Masterflex® pump
  • Choose from a wide range of power cords
PRODUCT OPTIONS
$18.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • AC Max (Volts)230
  • DescriptionPower Cord, 230 VAC, Australian Plug; 6-ft Long
Key Features
  • Choose from a wide range of power cords
More About this Item
Choose an interchangeable cord/plug assembly for 115 or 230 VAC operation. Each 6-ft (1.8 m) cord and moded plug assembly features an IEC 320 appliance connector and rated for 10 A at 250 VAC.