Masterflex Printed Microscope Slide, eight well; 144/pk
Representative image only.
Masterflex Item # EW-75955-27
Keep samples isolated
  • Screen-printed well design prevents bleed-through
  • Hydrophobic epoxy ink withstands frequent washing and resists chemicals
  • Contrasting ink makes orienting the microscope field of view over a sample easy
PRODUCT OPTIONS
$75.00USD / Pkg of 144
SPECS & DESCRIPTION
Specifications & Description
  • Length (")3
  • Slide TypePrinted with Mask
  • DescriptionPrinted Microscope Slide, eight well; 144/pk
Key Features
  • Screen-printed well design prevents bleed-through
  • Hydrophobic epoxy ink withstands frequent washing and resists chemicals
  • Contrasting ink makes orienting the microscope field of view over a sample easy
More About this Item
These printed microscope slides feature circular wells designed to hold samples in place, allowing multiple wells per slide, so you can increase throughput without adding to cleanup. Black PTFE-based epoxy makes locating wells with a microscope simple and quick, improving efficiency. Select from a range of well counts to suit any application.