Masterflex Printed Microscope Slide, one rectangular section; 144/pk
Representative image only.
Masterflex Item # EW-75955-33
Optimize sample testing and viewing
  • Screen-printed section design keeps samples contained
  • Hydrophobic epoxy ink withstands frequent washing and resists chemicals
  • Contrasting ink makes orienting the microscope field of view over a sample easy
PRODUCT OPTIONS
$68.50USD / Pkg of 144
SPECS & DESCRIPTION
Specifications & Description
  • Length (")3
  • Slide TypePrinted without Mask
  • DescriptionPrinted Microscope Slide, one rectangular section; 144/pk
Key Features
  • Screen-printed section design keeps samples contained
  • Hydrophobic epoxy ink withstands frequent washing and resists chemicals
  • Contrasting ink makes orienting the microscope field of view over a sample easy
More About this Item
Printed microscope slides are available with a variety of section shapes, sizes, counts, colors, and labels. The PTFE-based epoxy ink creates a hydrophobic barrier, holding samples in place for easy viewing under a microscope, and keeping reactions isolated from each other in multi-section slides.