Masterflex PTFE Check valve, 1/8" Orifice, 1/8" Female NPT
Representative image only.
Masterflex Item # EW-98515-12
Use with caustic and high-purity liquids in many applications
  • Constructed of durable PTFE
PRODUCT OPTIONS
$336.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.125
  • Outlet Port1/8" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/8" NPT(F)
  • Orifice (in)0.125
  • Orifice (mm)3.175
  • Max Pressure (PSI)50
  • Max Temperature (° F)212
  • Max Temperature (° C)100
  • DescriptionPTFE Check valve, 1/8" Orifice, 1/8" Female NPT
Key Features
  • Constructed of durable PTFE
More About this Item
Designed for use with corrosive fluids of varying viscosities in chemical, microelectronics, and petroleum industries.