Masterflex PTFE Multi-Turn Needle Valve, 1/8" NPT(F), 316 SS Body, 130 mL/min water
Representative image only.
Masterflex Item # EW-06393-80
Easily meter corrosive fluids or gases
PRODUCT OPTIONS
$79.00USD / Each
BUY MORE & SAVE
  • 1-3$79.00USD / Each
  • 4-9$71.10USD / Each
  • 10-19$67.15USD / Each
  • 20+$63.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.125
  • Outlet Port1/8" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/8" NPT(F)
  • Orifice (in)0.125
  • Orifice (mm)3.18
  • Max Flow Rate For Air (mL/min)2400
  • Max Flow Rate For Water (mL/min)130
  • Wetted MaterialsStainless Steel, PTFE, CTFE
  • DescriptionPTFE Multi-Turn Needle Valve, 1/8" NPT(F), 316 SS Body, 130 mL/min water
More About this Item
Maximum pressure is 75 psi; maximum temperature is 200°>F (93°>C). Wetted parts are body and O-ring made of PTFE and CTFE valve spindle. Choose aluminum (Al) or 316 stainless steel (SS) body. Orifice is 0.125".