Masterflex PVC Pressure Relief Valve, 1/4" NPT(F), 15 to 75 psig
Representative image only.
Masterflex Item # EW-00918-50
Easily meter fluids with pressure relief valves
  • Choose from different max pressures based on your application
$227.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.25
  • Outlet Port1/4" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/4" NPT(F)
  • Max Pressure (PSI)75
  • Tubing OD (in)1.375
  • Length (in)4.5
  • DescriptionPVC Pressure Relief Valve, 1/4" NPT(F), 15 to 75 psig
Key Features
  • Choose from different max pressures based on your application
More About this Item
These PVC valves have O-rings of Viton®, making them Ideal for liquid applications.