Masterflex QF1200-MF-SSSU MFLX QF1200 SS SINGLEUSE

Representative image only.
Masterflex Item # EW-74000-47
Moving delicate biological fluids—choose the four-chambered pump that was designed after the heart
  • Diaphragms enable soft, gentle pumping through the strokes generated by the eccentric shaft
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Ideal for cross-flow systems, tangential flow filtration (TFF), chromatography, cultures, and more
  • Choose from readily available single-use or multiple-use Quattroflow pump head systems

PRODUCT OPTIONS

$9,070.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Max Flow Rate (LPM)14
  • Min Flow Rate (LPM)0.33
  • Max Flow Rate (GPH)222
  • Min Flow Rate (GPH)5.3
  • Max Pressure (PSI)58
  • Max Pressure (bar)4
  • Motor (kW)0.26
  • Max Motor RPM (rpm)1700
  • Motor Enclosure TypeStainless steel
  • Wetted MaterialsPP (housing, valve plate), TPE (diaphragm), EPDM (valves, o-rings)
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Width (in)11.125
  • Length (in)16.75
  • Height (in)12.875
  • Width (cm)28.2
  • Length (cm)42.4
  • Height (cm)32.8
  • Motor hp3/8
  • DescriptionStainless Steel Single-use Pump System, 960 LPM
Two year warranty

Key Features

  • Diaphragms enable soft, gentle pumping through the strokes generated by the eccentric shaft
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Ideal for cross-flow systems, tangential flow filtration (TFF), chromatography, cultures, and more
  • Choose from readily available single-use or multiple-use Quattroflow pump head systems

More About this Item

The Quattroflow pump systems, featuring special Masterflex drives, are designed to safely and reliably pump delicate biologic fluids. The Quattroflow pump heads offer a unique four diaphragm (chamber) design, providing the constant low-pulsation, low-shear pumping needed for your biological fluids. In addition, the quaternary diaphragm design enables risk-free dry-running, self-priming, and a high turn-down ratio. Both the single-use and multiple-use models safely and reliably pump plasma products, proteins, monoclonal antibodies, vaccines, and more.

Single-use pump heads have a disposable wetted chamber constructed of solid polypropylene (PP) that can be replaced as a complete unit. This saves you time and money by eliminating the cleaning validation, sterilization process normally associated with permanent multiple-use systems. You can quickly turn around equipment from one batch to the next, maximizing production operations. Plus, fluid path integrity and sterility is ensured with no chance of cross-batch or cross-product contamination. Come presterilized and double bagged. It is recommended you replace the pump head every 1000 hours of operation.

Multiple-use pump heads ensure total product containment without abrasion and minimal particle creation. Stainless steel construction handles higher pressure applications including cross-flow filtration and chromatography columns.

The special Masterflex drives, enclosed in ABS plastic or stainless steel, were developed to best take advantage of the operating parameters of the Quattroflow pumps. They feature an intuitive interface, remote control functionality, and a brushless, maintenance-free motor with ±0.1% speed control accuracy. The graphical LCD shows six different parameters: motor rpm, flow rate, dispense volume, cumulative volume, dispense interval time, and copy number. Dispense by volume or time with a user-defined time interval between cycles.

The plastic-enclosed drives have an IP33 rating for benchtop lab applications and have been combined with the lower flow QF150 series pump head.
Stainless steel drives have IP66 and NEMA 4X ratings and are designed for both the QF150 and QF1200 series. Sealed stainless steel housing and sealed keypad are protected from dust, water, and chemicals in the washdown environment—simply hose down to clean.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat