Masterflex Quattroflow QF1200S Mutiple-Use Pump Chamber; 316L machined

Representative image only.
Masterflex Item # EW-76000-08
Keep your Quattroflow™ pump system up and running with these quality replacement pump heads
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Single-use pump heads are constructed with a disposable polypropylene chamber
  • Reusable pump heads are constructed of durable 316L stainless steel
  • Ideal replacement for stainless steel multiuse Quattroflow pump systems

PRODUCT OPTIONS

$3,322.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Wetted Materials316L SS (housing, valve plate), TPE (diaphragm), EPDM (valves, O-rings)
  • Inlet Connection3⁄4" Tri-Clamp
  • Outlet Connection3⁄4" Tri-Clamp
  • DescriptionMutiple-Use Pump Chamber; 316L machined stainless steel multiuse Quattroflow pump systems

Key Features

  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Single-use pump heads are constructed with a disposable polypropylene chamber
  • Reusable pump heads are constructed of durable 316L stainless steel
  • Ideal replacement for stainless steel multiuse Quattroflow pump systems

More About this Item

Keep your Quattroflow pump system up and running with these quality replacement pump heads. The quaternary diaphragm design enables risk-free dry-running, self-priming, and a high turn-down ratio. These pump heads can safely and reliably pump plasma products, proteins, monoclonal antibodies, vaccines, and more.

Order the Masterflex® Single-use Quattroflow™ Pump Systems separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat