Masterflex Replacement Tubing Set, Norprene®, 0.2 to 44.4 GPH Flow Range
Representative image only.
Masterflex Item # EW-74204-42
Choose tubing based on chemical compatibility, flow rate, and your current pump
  • Suitable for use with your Masterflex® High-Pressure Peristaltic Pump
PRODUCT OPTIONS
$119.30USD / Set of 1
SPECS & DESCRIPTION
Specifications & Description
  • Max Flow Rate (mL/min)2800
  • Min Flow Rate (mL/min)1.1
  • Max Flow Rate (LPH)168
  • Max Pressure (PSI)125
  • Suction Lift, Wet (in H₂O)360
  • Suction Lift, Wet (mm of H₂O)9144
  • Max Motor RPM (rpm)125
  • Wetted MaterialsLLDPE, Norprene, PVC, PVDF
  • Inlet Connection1/2" hose barb
  • Outlet Connection1/2" hose barb
  • Max Temperature (° F)185
  • Max Temperature (° C)85
  • Viscosity, Max (cP)12000
  • DescriptionReplacement Tubing Set, Norprene®, 0.2 to 44.4 GPH Flow Range
Key Features
  • Suitable for use with your Masterflex® High-Pressure Peristaltic Pump
More About this Item
Choose the tubing that is compatible with your pump and meets your application needs.