Masterflex Sanitary Clamp with White Knob, Nylon, 1/2" to 3/4"; 1/EA
Representative image only.
Masterflex Item # EW-30508-21
Compatible with metal and plastic Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics
  • Autoclavable
  • Accepts 1/2" to 3/4" tubing
PRODUCT OPTIONS
Free Certificate of Comliance
$26.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialNylon
  • Sterilization MethodAutoclave or Ethylene oxide (ETO)
  • Connection TypeSanitary
  • Sanitary Clamp Size (Port 1) (in)3/4
  • DescriptionNylon Clamp for Sanitary Fittings, 1/2" - 3/4"
Certificate of Compliance
Key Features
  • Excellent chemical resistance; low leachable characteristics
  • Autoclavable
  • Accepts 1/2" to 3/4" tubing
More About this Item
Constructed of nylon, these Tri-Clamp® sanitary clamps are perfect for single-use transfer lines in biopharma markets, plus a wide range of fluid transfer systems. These clamps ideal for securing your fluid transfer system without damaging the process lines.