This item is now discontinued. Please browse similar items
Masterflex Satellite cable; RJ11 to RJ11 connectors; for daisy chaining drive to drives or Servodyne mixers,
Representative image only.
Masterflex Item # EW-07550-66
Easily connect your Masterflex® L/S® computer-compatible drive to a PC
  • Featuring RS-232 connections with 8 foot cable length
SPECS & DESCRIPTION
Specifications & Description
  • Operating PlatformPC
  • DescriptionSatellite cable; RJ11 to RJ11 connectors; for daisy chaining drive to drives or Servodyne mixers,
Key Features
  • Featuring RS-232 connections with 8 foot cable length
More About this Item
Monitor and control your Servodyne mixer system from an IBM® or 100% compatible computer via a computer interface. Software programs let you to control up to 16 Servodyne mixers and Masterflex® computer-compatible pumps. Various application programs let you dedicate your automated setup for mixing and diluting, sequence control, flow proportioning, reversible batch dispensing, delivering single and multiple slope gradients, or customizing procedures for specialized applications.