Masterflex Single Pump Wall Mount System Kit; 1/Each
Representative image only.
Masterflex Item # EW-70791-31
Cost-effective, unassembled kits for your pH metering applications
  • Single- and dual-pump configurations
  • Purchase pump(s) separately or use your existing metering pumps
$1,866.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Wetted MaterialsPVC, Viton
  • DescriptionSystems, Single Pump Wall Mount System, Ready for Assembly
Includes
Chemical Suction Inlet: 1/2" PVC ball valve, ½” PVC fitting, 100 mL PVC calibration column, 1/2" Isolation ball valve, Calibration Inlet: 1/2” PVC ball valve, pressure gauge with Viton® gauge guard, ½” NPT PVC coupling, pipe supports, 1/2" PVC Pipe, Chemical Discharge Outlet: 1/2" PVC Ball Valve, Pump Shelf
Key Features
  • Single- and dual-pump configurations
  • Purchase pump(s) separately or use your existing metering pumps
More About this Item
Any brand of metering pump can be used with these packages