Masterflex Single-Use Quattroflow™ Disposable Pump Chamber with EZ-Set, Polypropylene, 3/4" Tri-Clamp
Representative image only.
Masterflex Item # EW-76000-03
Keep your Quattroflow™ pump system up and running with these quality replacement pump heads
  • EZ-Set design allows for quick pump head change—reducing downtime
  • Constructed with a disposable polypropylene chamber—ideal for single-use applications
  • Diaphragms enable soft, gentle pumping through the strokes generated by the eccentric shaft
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Replacement single-use pump head for the 14 LPM Single-use Quattroflow™ Pump System 74000-59
PRODUCT OPTIONS
$1,851.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • SeriesQuattroflow
  • Max Flow Rate (LPM)14
  • Min Flow Rate (LPM)0.33
  • Max Flow Rate (PSI)58
  • Wetted MaterialsPolypropylene (housing, valve plate), TPE (diaphragm), EPDM(valves, O-rings)
  • Inlet Connection3⁄4" Tri-Clamp
  • Outlet Connection3⁄4" Tri-Clamp
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • DescriptionDisposable Pump Chamber for Pump System 74000-59
Key Features
  • EZ-Set design allows for quick pump head change—reducing downtime
  • Constructed with a disposable polypropylene chamber—ideal for single-use applications
  • Diaphragms enable soft, gentle pumping through the strokes generated by the eccentric shaft
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Replacement single-use pump head for the 14 LPM Single-use Quattroflow™ Pump System 74000-59
More About this Item
The Masterflex® Quattroflow™ single-use replacement pump heads offer a unique four diaphragm (chamber) design, providing the constant low-pulsation, low-shear pumping needed for your biological fluids. In addition, the quaternary diaphragm design enables risk-free dry-running, self-priming, and a high turn-down ratio. These pump heads can safely and reliably pump plasma products, proteins, monoclonal antibodies, vaccines, and more.
Single-use pump heads have a disposable wetted chamber constructed of solid polypropylene (PP) that can be replaced as a complete unit. This saves you time and money by eliminating the cleaning validation/sterilization process. Plus, fluid path integrity is ensured with no chance of cross-batch or cross-product contamination. These single-use pump heads can be sterilized via autoclave or gamma irradiation. With the EZ-Set pump head design, you can change your single-use pump chamber in 30 seconds without the use of special tools or torque wrenches.
Order the Masterflex® Single-use Quattroflow™ Pump Systems separately.