Masterflex Slide Forceps, plastic, wide tip; 50/pk
Representative image only.
Masterflex Item # EW-75955-11
No more cross-contamination
  • Protect yourself and your samples with disposable forceps
  • Autoclavable plastic or stainless steel construction
PRODUCT OPTIONS
$96.00USD / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)4 1/2
  • Overall Length (cm)11.4
  • MaterialPlastic
  • SterileNo
  • DescriptionSlide Forceps, plastic, wide tip; 50/pk
Key Features
  • Protect yourself and your samples with disposable forceps
  • Autoclavable plastic or stainless steel construction
More About this Item
These single-use forceps are bulk-packed for convenience and affordability. Slide forceps feature an extra-wide grip for greater stability and security when staining cells and tissue. Fine-tipped forceps are ideally suited for work with smaller specimens. Stainless steel forceps have grooved finger grips and ridged tips, best for larger, soft or wet specimens.