Masterflex Solenoid-Operated Three-Way Pinch Valve, 12 VDC
Representative image only.
Masterflex Item # EW-98305-00
Excellent choice for viscous fluids
PRODUCT OPTIONS
$153.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Pinch Valve Tube Size2.4 mm OD
  • Connection TypePinch Valve for Tubing
  • Power (VDC)12
  • Watts4.4
  • Flow PatternThree-Way, One Normally Closed, One Normally Open
  • DescriptionSolenoid-Operated Three-Way Pinch Valve, 12 VDC
More About this Item
These solenoid-operated 2- and 3-Way pinch valves are ideal for use with fluids that are difficult for traditional solenoid valves to handle, such as viscous fluids. They are ideal for on/off flow control applications and where frequent changes of wetted parts are necessary. All models are compatible with silicone, C-FLEX®, and PharMed® BPT tubing. The two-way valves are available in both normally closed and normally open configurations. The three-way valves accept two tubes, the bottom tube is normally open while the top is normally closed.