Masterflex Turbine Flow Sensor, 316L Stainless Steel, 0.5 to 20 LPM, 3/8" NPT(M), with Single-Point Calibration Certificate
Representative image only.
Masterflex Item # EW-24507-39
Build a complete flow sensor system customizable to your application
  • Use in conjunction with batch or flow controller for precise pump control
  • Accurate 1% of reading measurement at low flow
  • Flow sensors are produced and tested under cleanroom conditions
  • Capable of 0.5 to 20 LPM flow rate
PRODUCT OPTIONS
$545.00USD / Each
BUY MORE & SAVE
  • 1-4$545.00USD / Each
  • 5-7$490.50USD / Each
  • 8-9$463.25USD / Each
  • 10+$436.00USD / Each
INCLUDE TODAY…
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy1% of reading
  • Repeatability<0.15%
  • Max Pressure (PSI)2900
  • Min Flow Rate (LPM)0.5
  • Max Flow Rate (LPM)20
  • Material Fitting316 Stainless Steel
  • Material O-RingEPDM
  • Material FloatRuby
  • Material Housing316 Stainless Steel and PVDF
  • Wetted Materials316 SS, PVDF, and ruby sensor with EPDM o-ring
  • Min Operating Temperature (° F)-4
  • Min Operating Temperature (° C)-20
  • Max Operating Temperature (° F)176
  • Max Operating Temperature (° C)80
  • Process Connection (NPT(M))0
  • Process Connection3/8" NPT(M)
  • Fluid TypeConductive or non-conductive fluids with no lenticular impurities
  • Viscosity0.8 to 10 cSt
  • Power5 to 24 VDC
  • OutputOne 5 to 24 volt square wave pulse
  • CertifiedSingle-Point Calibration with Certificate
  • DescriptionTurbine Flow Sensor, 316L Stainless Steel, 0.5 to 20 LPM, 3/8" NPT(M), with Single-Point Calibration Certificate
Includes
single-point calibration certificate.
Key Features
  • Use in conjunction with batch or flow controller for precise pump control
  • Accurate 1% of reading measurement at low flow
  • Flow sensors are produced and tested under cleanroom conditions
  • Capable of 0.5 to 20 LPM flow rate
More About this Item
The Masterflex® stainless steel (SS) turbine flow sensors are corrosion resistant for long-term, trouble-free performance in demanding situations. These are rated for high-pressure requirements in chemical, pharmaceutical, semiconductor, and environmental industries.
The sensor is positioned in close proximity to the edges of the rotor blades. In conjunction with the ultra-lightweight turbine rotor, minor fluctuations in flow can accurately be measured for a high-resolution flow measurement for liquids. Flow of the liquid is picked up via the IR beam in the sensor connection and converted into a 5 to 24 V square wave pulse output. Mount these turbine flow sensors in any position; they do not require a flow straightener or other specialized installation. All flow sensors come precalibrated to either one or five points. All single-point calibrated flow sensors come with a calibration certificate with a K-factor at 50% of the flowrate and serial number. All 5-point calibrated flow sensors come with a calibration certificate with a K-factor at 5 points of flowrate and serial number.
Four output options are available — digital to analog converter, metering pump control system, panel mount flow display with totalizer, and benchtop flow display with totalizer. Order barcode scanner and output option separately.