Masterflex USB to DB9 (M) RS-232 Serial Adapter Cable
Representative image only.
Masterflex Item # EW-22050-58
$49.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionUSB to DB9 (M) RS-232 Serial Adapter Cable
More About this Item
Use this cable to connect the Masterflex Computer Compatible drives 07550- and 07551- series to a PC with USB port. Will also connect other peripheral devices equipped with RS-232 serial interface including balances, printers, scanners, and PDA's. Includes 2-ft cable and driver software. Supports data transfer speeds up to 12 Mbps; compatible with Windows® 2000 / XP / Vista / 7.