Masterflex Variable-Area Flowmeter, Correlated Reading, Dual-Scale, Polycarbonate Shield; 137,000 mL/min Air
Representative image only.
Masterflex Item # EW-03205-20
Don’t sacrifice suitability for precision
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Dual-unit scales for air or water offer flexibility
PRODUCT OPTIONS
$621.00USD / Each
$272.00USD / Each
for an additional
$272.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy0.05% of Correlated reading
  • Max Pressure (PSI)50
  • Max Air Flow Rate (mL/min)137000
  • Max Water Flow Rate (mL/min)4100
  • Min Flow Rate (mL/min)3000
  • Max Flow Rate (mL/min)4100
  • Material FloatGlass or 316 SS
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Pipe Size5
  • Fluid TypeWater
  • Gas TypeAir
  • Media TypeAir
  • DescriptionShielded Correlated Flowmeter, Tube Size 5
Key Features
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Dual-unit scales for air or water offer flexibility
More About this Item
Extremely accurate—±2% of reading or ±1 scale division, whichever is greater. Flowmeters with micro-sized tubes (models 03210-00, 03210-20, and 03234-50) are accurate to ±5% of reading or ±2% of scale whichever is greater. Computer calibrated flow charts are included for floats used with both air and water at STP. An “R factor” chart and formulas are included to convert scale readings for other gases or liquids or for floats other than glass. Order our flow analysis software below to generate flow charts specific to your individual applications.
Shielded models accept 3/8" ID tubing (sizes 4 and 5 accept 5/8" ID tubing). PTFE coupling adapters for unshielded models are not included—order separately.