Masterflex Variable-Area Flowmeter, Correlated Reading, PTFE Housing and Fittings, 150-mm Scale; 825 mL/min Air
Representative image only.
Masterflex Item # EW-03267-67
Don’t sacrifice suitability for precision
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
PRODUCT OPTIONS
$337.80USD / Each
BUY MORE & SAVE
  • 1-3$337.80USD / Each
  • 4-9$304.02USD / Each
  • 10-19$287.13USD / Each
  • 20+$270.24USD / Each
$291.00USD / Each
for an additional
$291.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Repeatability±0.25% full-scale
  • Accuracy±2% full-scale
  • Max Pressure (PSI)100
  • Max Air Flow Rate (mL/min)825
  • Max Argon Flow Rate (mL/min)687
  • Max CO2 Flow Rate (mL/min)725
  • Max H2 Flow Rate (mL/min)2620
  • Max Water Flow Rate (mL/min)16.5
  • Max Helium Flow Rate (mL/min)1490
  • Max N2 Flow Rate (mL/min)827
  • Max O2 Flow Rate (mL/min)772
  • Max Flow Rate (mL/min)825
  • Material Flow TubeBorosilicate glass
  • Material FittingPTFE and Kel-F®
  • Material ValvePTFE and Kel-F®
  • Material O-RingPTFE
  • Material FloatGlass
  • Material HousingAluminum
  • ValveNo
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Process Connection (NPT(F))0
  • Description150-mm Correlated Flowmeter, w/o valve, PTFE, 825 mL/min air
Key Features
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
More About this Item
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. PTFE has the best chemical compatibility.