Masterflex Variable-Area Flowmeter, Direct Read, Acrylic Housing, 100 mm Scale; 0.4 to 5 LPM Air
Representative image only.
Masterflex Item # EW-32458-50
Durable single-piece body withstands severe shock and vibration
  • Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
  • Flexible design allows for panel or bench mounting
  • Variable Area with Direct Metric-Reading Scales
  • Maximum flow rate of 5 LPM
PRODUCT OPTIONS
$125.50USD / Each
BUY MORE & SAVE
  • 1-3$125.50USD / Each
  • 4-9$112.95USD / Each
  • 10-19$106.68USD / Each
  • 20+$100.40USD / Each
$291.00USD / Each
for an additional
$291.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±3% full-scale
  • Repeatability±0.5% full-scale
  • Max Pressure (PSI)100
  • Min Flow Rate (mL/min)400
  • Max Flow Rate (mL/min)5000
  • Material FittingBrass
  • Material O-RingBuna-N
  • Material Float316 Stainless Steel
  • Material HousingAcrylic
  • ValveNo
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Process Connection (NPT(F))0
  • Media TypeAir
  • DescriptionVariable-Area Flowmeter, Direct Reading, Acrylic Housing, 100 mm Scale; 0.4 to 5 LPM Air
Key Features
  • Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
  • Flexible design allows for panel or bench mounting
  • Variable Area with Direct Metric-Reading Scales
  • Maximum flow rate of 5 LPM
More About this Item
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters’ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately below) which allows for mobility from bench to bench.
Note: There are many additional types of acrylic flowmeters not listed here. Contact our Application Specialists for quotes on acrylic meters with special requirements.