Masterflex Variable-Area Flowmeter, Direct Read, Acrylic Housing, 100 mm Scale; 3 to 30 LPM Air
Representative image only.
Masterflex Item # EW-32458-56
Durable single-piece body withstands severe shock and vibration
  • Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
  • Flexible design allows for panel or bench mounting
  • Variable Area with Direct Metric-Reading Scales
  • Maximum flow rate of 30 LPM
PRODUCT OPTIONS
$144.80USD / Each
BUY MORE & SAVE
  • 1-3$144.80USD / Each
  • 4-9$130.32USD / Each
  • 10-19$123.08USD / Each
  • 20+$115.84USD / Each
$291.00USD / Each
for an additional
$291.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±3% full-scale
  • Repeatability±0.5% full-scale
  • Max Pressure (PSI)100
  • Min Flow Rate (mL/min)3000
  • Max Flow Rate (mL/min)30000
  • Material FittingBrass
  • Material O-RingBuna-N
  • Material Float316 Stainless Steel
  • Material HousingAcrylic
  • ValveNo
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Process Connection (NPT(F))0
  • Media TypeAir
  • DescriptionVariable-Area Flowmeter, Direct Reading, Acrylic Housing, 100 mm Scale; 3 to 30 LPM Air
Key Features
  • Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
  • Flexible design allows for panel or bench mounting
  • Variable Area with Direct Metric-Reading Scales
  • Maximum flow rate of 30 LPM
More About this Item
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters’ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately below) which allows for mobility from bench to bench.
Note: There are many additional types of acrylic flowmeters not listed here. Contact our Application Specialists for quotes on acrylic meters with special requirements.