Masterflex Variable-Area Flowmeter, Direct Reading, Acrylic Housing and Brass Fittings without valve, 3.5 L/min Oxygen
Representative image only.
Masterflex Item # EW-68560-30
Designed for labs and plant processes where multiple gases are used
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
PRODUCT OPTIONS
$53.15USD / Each
BUY MORE & SAVE
  • 1-3$53.15USD / Each
  • 4-9$47.84USD / Each
  • 10-19$45.18USD / Each
  • 20+$42.52USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5% full-scale
  • Max Pressure (PSI)100
  • Max Flow Rate (LPM)3.5
  • Material FittingBrass
  • Material O-RingViton®
  • Material HousingAcrylic
  • ValveNo
  • Wetted MaterialsBrass
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Media TypeOxygen
  • DescriptionAcrylic Flowmeter w/o Valve for O2, Brass, 3.5 LPM
Key Features
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
  • Flowmeter features brass fittings and Viton® o-ring
More About this Item
Versatile acrylic flowmeters are designed specifically for air, oxygen, nitrogen, carbon dioxide, argon, helium, or water. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. Flowmeters feature a stable, easy-to-read float for quick measurement readings.