Masterflex Variable-Area Flowmeter, Direct Reading, Dual-Scale, Polycarbonate Shield; 3700 mL/min Air
Representative image only.
Masterflex Item # EW-03232-22
Design makes each meter suitable for a broad range of laboratory applications
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate│Dual-unit scales for air or water offer flexibility
  • Features a 1/4" NPT(M) port and accepts 3⁄8" ID tubing
  • Maximum pressure range of 100 psi
PRODUCT OPTIONS
$304.00USD / Each
$272.00USD / Each
for an additional
$272.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5% of reading or 2 mm of the scale length, whichever is greater
  • Max Air Flow Rate (mL/min)3700
  • Max Water Flow Rate (mL/min)86
  • Max Flow Rate (mL/min)3700
  • Material Flow TubeBorosilicate glass
  • Material FittingPTFE
  • Material ValveGlass chamber with PCTFE plug
  • Material O-RingViton®
  • Material FloatGlass or 316 SS
  • Material HousingPC
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Fluid Typeair and water
  • Media TypeAir
  • Gas TypeAir
  • Display TypeSize 12 Tube
  • DescriptionVariable Area Shielded Flowmeters, size 12
Key Features
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate│Dual-unit scales for air or water offer flexibility
  • Features a 1/4" NPT(M) port and accepts 3⁄8" ID tubing
  • Maximum pressure range of 100 psi
More About this Item
Choose from unshielded, shielded, or shielded with valve flow meters suitable for a wide range of applications.
Unshielded Flowmeters work well in low-pressure applications. Connect the flowtube end directly to tubing having the proper inner diameter. Maximum pressure rating for unshielded flowmeters is 15 psi.
Shielded Flowmeters are better suited to higher-pressure applications or for installations requiring a panel-mounted flowmeter. End bushings are molded with dual connection capability—tubing or threaded.
Shielded Flowmeters with Valves offer flow monitoring along with precise control through an integrated 20-turn micrometer valve. The valve can be adjusted from 0.1 to 100% of maximum flow (semilogarithmic) and provides recise regulation from 0.3 to 60% of maximum flow.
Glass taper joints are available upon request—call our Application Specialists to order.