Masterflex Variable-Area Flowmeter, Direct-Reading, Stainless Steel Housing, Tube Size 5; 3.8 GPM
Representative image only.
Masterflex Item # EW-32447-34
Ideal for the most aggressive process applications
  • Thick polycarbonate front shield provides protection
  • Unique design facilitates ease of maintenance
  • Easy-to-read direct-reading scale offers both metric and English measurements
  • Available in fluted or plain tapered tubes
PRODUCT OPTIONS
$644.50USD / Each
BUY MORE & SAVE
  • 1-3$644.50USD / Each
  • 4-9$580.05USD / Each
  • 10-19$547.83USD / Each
  • 20+$515.60USD / Each
$272.00USD / Each
for an additional
$272.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±3% full-scale
  • Repeatability±0.5% full-scale
  • Max Flow Rate (GPM)3.8
  • Material Flow TubeBorosilicate glass
  • Material Fitting316 SS
  • Material O-RingViton®
  • Material Float316 SS
  • Material Housing304 SS
  • Max Operating Temperature (° F)200
  • Max Operating Temperature (° C)93
  • Pressure Drop At Maximum Flow (water column)10
  • DescriptionSS Shielded Direct-Reading Flowmeter, 3.8 GPM, Tube Size 5
Key Features
  • Thick polycarbonate front shield provides protection
  • Unique design facilitates ease of maintenance
  • Easy-to-read direct-reading scale offers both metric and English measurements
  • Available in fluted or plain tapered tubes
More About this Item
Detachable, clear 3/16"-thick polycarbonate front shield provides protection at maximum rated temperature and pressure. Easily read the graduated direct-reading scale. The interchangeable transparent scale plates mount directly on the front of the flowtube. Unique float stops allow easy cleaning-remove and replace the float without disturbing either the flowtube or the shield. Tube sizes 3, 4, and 5 are fluted; tube sizes 6, 8, and 9 are tapered with a stainless steel guide rod.