Masterflex Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame System, Range 1
Representative image only.
Masterflex Item # EW-03218-72
Control your flow with the addition of a valve
  • Aluminum is economical and good for general use with noncorrosive gases and liquids
  • 316 Stainless steel (SS) withstands higher temperatures and pressures, and features excellent chemical compatibility
PRODUCT OPTIONS
$82.20USD / Each
BUY MORE & SAVE
  • 1-3$82.20USD / Each
  • 4-9$73.98USD / Each
  • 10-19$69.87USD / Each
  • 20+$65.76USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Width (")1754
  • Min Air Flow Rate (mL/min)0
  • Max Air Flow Rate (mL/min)200
  • Min Water Flow Rate (mL/min)0
  • Max Water Flow Rate (mL/min)6
  • Max Flow Rate (mL/min)200
  • Material O-RingBuna-N
  • Material HousingAluminum
  • DescriptionVariable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 1
Key Features
  • Aluminum is economical and good for general use with noncorrosive gases and liquids
  • 316 Stainless steel (SS) withstands higher temperatures and pressures, and features excellent chemical compatibility
More About this Item
Select a valve-cartridge that matches your frame. Insert these needle valves into the flowmeter frames to control flow rate. Choose a 16-turn, high-resolution metering valve-cartridge for greater precision of flow rate. If your flowtube is in units other than mL/min, convert them to mL/min and choose a valve within that flow range from the table below. See website and virtual catalog page for more details.