Masterflex Variable-Area Flowmeter with High-Res Valve, Correlated Reading, 316 SS Housing and Fittings, 65-mm Scale, 2520 mL/min Air
Representative image only.
Masterflex Item # EW-32046-19
Don’t sacrifice suitability for precision
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
PRODUCT OPTIONS
$405.00USD / Each
BUY MORE & SAVE
  • 1-3$405.00USD / Each
  • 4-9$364.50USD / Each
  • 10-19$344.25USD / Each
  • 20+$324.00USD / Each
$291.00USD / Each
for an additional
$291.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±2% full-scale
  • Repeatability±0.25%
  • Max Flow Rate (mL/min)70.7
  • Material Flow TubeBorosilicate glass
  • Material Fitting316 SS
  • Material Valve316 SS
  • Material O-RingViton®
  • Material Float316 SS
  • Material Housing316 SS
  • Max Operating Temperature (° F)250
  • Max Operating Temperature (° C)121
  • Process Connection (NPT(F))0
  • Fluid Typewater
  • Description65-mm Correlated Flowmeter, hi-res valve, 316 SS, 2520 mL/min air
Key Features
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
More About this Item
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).