Masterflex Variable-Area Flowmeter without Valve, Direct-Reading, Aluminum Fittings, 150-mm Scale; 10 L/min Air
Representative image only.
Masterflex Item # EW-32000-14
No more eye strain—front shield magnifies scale 16 percent for easier readings
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
PRODUCT OPTIONS
$183.50USD / Each
BUY MORE & SAVE
  • 1-3$183.50USD / Each
  • 4-9$165.15USD / Each
  • 10-19$155.98USD / Each
  • 20+$146.80USD / Each
$291.00USD / Each
for an additional
$291.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5% full-scale
  • Repeatability±0.25% full-scale
  • Max Flow Rate (LPM)10
  • Max Flow Rate (mL/min)10000
  • Material Flow TubeBorosilicate glass
  • Material FittingAluminum
  • Material O-RingBuna-N
  • Material FloatGlass
  • Material HousingAluminum
  • ValveNo
  • Max Operating Temperature (° F)250
  • Max Operating Temperature (° C)121
  • Process Connection (NPT(F))0
  • Fluid TypeAir
  • Description150-mm Direct Reading Flowmeter, Al/Gl Float; Air, 10 LPM
Includes
150-mesh inlet screen.
Key Features
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
  • Aluminum is economical and good for general use with noncorrosive gases and liquid
  • Glass float is black and has a density of 2.53 g/mL
  • Max flow for air is 10 LPM
More About this Item
This full range of direct-reading flowmeters provide a robust solution for your laboratory that eliminates the need for reading conversions. Valved flowmeters have a valve mounted at the inlet (bottom) of the flowmeter. This arrangement is typically used for positive pressure applications for liquids and gases. The valve can be reconfigured to mount at the outlet (top), often for vacuum applications.