Masterflex Variable-Area Flowmeter without Valve, Direct Reading, PTFE Fittings, 65-mm Scale; 35 std mL/min Hydrogen
Representative image only.
Masterflex Item # EW-32007-24
Ideal for gas applications where metal components must be minimized
  • Transparent front safety shield enhances reading resolution by 16%
  • Rugged design features case-enclosed flowtube
  • Tubes include a fused ceramic scale as a precise, permanent measuring guide
PRODUCT OPTIONS
$249.60USD / Each
BUY MORE & SAVE
  • 1-3$249.60USD / Each
  • 4-9$224.64USD / Each
  • 10-19$212.16USD / Each
  • 20+$199.68USD / Each
$272.00USD / Each
for an additional
$272.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Repeatability±0.25% full-scale
  • Accuracy±5% full-scale
  • Material Flow TubeBorosilicate glass
  • Material FittingPTFE
  • Material ValvePTFE
  • Material O-RingPTFE
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Process Connection (NPT(F))0
  • Media TypeHydrogen
  • Gas TypeHydrogen
  • DescriptionDirect Reading PTFE-Glass Flowmeter, 65-mm, , Hydrogen, 35 std mL/min
Key Features
  • Transparent front safety shield enhances reading resolution by 16%
  • Rugged design features case-enclosed flowtube
  • Tubes include a fused ceramic scale as a precise, permanent measuring guide
More About this Item
These meters eliminate the need for correlation charts with direct-reading scales for individual routine gases. The inert PTFE and glass wetted parts combine with the rigid anodized aluminum frame to offer a unique solution for ultra-pure gas applications. Flowtubes are replaceable.