Masterflex Variable-Area Single-Tube Frame, 65-mm Scale; No Valve Port, Aluminum
Representative image only.
Masterflex Item # EW-03220-00
Create your own flowmeter systems for complete flexibility
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Use with media for laboratory and industrial applications
  • Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
  • Choose from aluminum or stainless steel wetted parts based on required chemical compatibility
PRODUCT OPTIONS
$58.15USD / Each
BUY MORE & SAVE
  • 1-3$58.15USD / Each
  • 4-9$52.34USD / Each
  • 10-19$49.43USD / Each
  • 20+$46.52USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure (PSI)200
  • Material HousingAluminum
  • ValveNo Valve Port
  • Max Operating Temperature (° F)250
  • Max Operating Temperature (° C)121
  • Process Connection (NPT(F))0
  • DescriptionVariable-Area Single-Tube Frame, 65-mm Scale; No Valve Port, Aluminum
Key Features
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Use with media for laboratory and industrial applications
  • Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
  • Choose from aluminum or stainless steel wetted parts based on required chemical compatibility
  • Designed with 1/8” NPT(F) connections
More About this Item
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs.
Choose a frame depending on the flowtube capacity, flow pattern, and wetted parts that you require.
Choose a frame with a valve port if you plan on adding a valve cartridge to control flow.