Masterflex Versatile Cap Adapter Insert, PP, 50 mm, vent and 2 x 5/16" ID Hose Barb ports; 1 ea.
Representative image only.
Masterflex Item # EW-06058-69
Interchangeable ports for flexibility
  • Choose from a variety of sizes and configurations
  • Change the top connections of each carboy to match your application needs
  • Autoclavable
PRODUCT OPTIONS
$30.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialPP
  • Cap / Stopper Size53B
  • Cap ColorWhite
  • AutoclavableYes
  • Tubing IDVent and 2 x 5/16" ID Hosebarb Ports
  • DescriptionVersatile Cap Adapter Insert, 53B, Vent and 2 x 5/16" ID Hosebarb Ports
Key Features
  • Choose from a variety of sizes and configurations
  • Change the top connections of each carboy to match your application needs
  • Autoclavable
More About this Item
Versatile carboy cap adapter inserts are made of autoclavable polypropylene or clear polycarbonate. Quick-connect hosebarb fittings are also available. Vent ports are 1/4" in diameter.