McMillan Flow 210R-I Flow Rate Indicator with miniature 3-1/2 Digit LCD Display
Representative image only.
McMillan Flow Mfr # 210R-I – Item # EW-32706-72
$210.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Input Signal0 to 5 VDC
  • Display TypeMiniature 3-1/2 Digit LCD Indicator
  • DescriptionFlow Rate Indicator with miniature 3-1/2 Digit LCD Display
More About this Item
Select a flow rate/total display for use with flowmeters / transmitters. Note the signal-input requirements for each.
Contact our Application Specialists for correct pairing.
This low-power device operates from and existing 5 to 24 VDC source. View the 0.4" high digits in any flow unit combination to which the transmitter output signal is scaled. Panel cutout is 1.67"L x 0.92"H (x 1.00"D). The input signal is 0 to 5 VDC.