Mecmesin 870-002-V03 Smart "S" Type Load Cell Tranducer/Sensor; 22 lb
Representative image only.
Mecmesin Mfr # 870-002-V03 – Item # EW-59887-61
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)22
  • Capacity (Newtons)100
  • Accuracy±0.25% of full scale
  • Overload120% of full scale
  • DescriptionSmart \"S\" Type Load Cell Tranducer/Sensor; 22 lb
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Use the "S" type load cells where space is not limited; dedicated fixtures attach via threaded holes. Miniature button cell is used for compression only, and is ideal for applications in restricted spaces. Static torque screwdrivers may be handheld or mounted.