Megger 1012-069 Grabber Clip Lead Set, CAT III 100 V, CAT IV 600 V; for MTR105

Representative image only.
Megger Mfr # 1012-069 – Item # EW-20026-60
Optimize performance of your Megger rotating machine tester
  • Compatible with Megger MTR105 Rotating Machine Tester (20026-56)
  • CAT III 100 V & CAT IV 600 V rated
  • Red, blue, and black
  • 1 year warranty

PRODUCT OPTIONS

$66.00USD / Each
sign in for your price.

SPECS & DESCRIPTION

Specifications & Description

  • Safety RatingCAT III 100 V
  • DescriptionGrabber Clip Lead Set, CAT III 100 V, CAT IV 600 V; for MTR105
  • Warranty1 year
1 Year Warranty

Key Features

  • Compatible with Megger MTR105 Rotating Machine Tester (20026-56)
  • CAT III 100 V & CAT IV 600 V rated
  • Red, blue, and black
  • 1 year warranty

More About this Item

Choose from the following line of Megger rotating machine tester accessories.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat