Megger 220005 5 kV DC Dielectric Test Set
Representative image only.
Megger Mfr # 220005 – Item # EW-20022-66
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DC Max (Volts)5
  • AC Max (Volts)5
  • Description5 kV DC Dielectric Test Set
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The Dielectric Test Sets measure leakage current while applying a dc voltage at or above the insulation system’s operating level. This measurement aids in determining the insulation system’s ability to withstand overvoltages such as lightning strikes and switching surges.
This portable DC dielectric test sets check the electrical insulation quality of motors, power cables, switchgear, insulators, transformers and capacitors. Typical applications include acceptance and maintenance testing of critical equipment used by electrical utility substations and industrial plant distribution systems. Power apparatus manufacturers may also use the equipment to perform QA/QC production tests.
The test sets can be used to perform step-voltage and proof tests which, when incorporated into a routine maintenance program, can aid in predicting potential failure before breakdown occurs.
FEATURES
  • Variable test voltage
  • Rugged field construction
  • Compact and portable
  • Line powered 12 VAC, 50/60 Hz