Megger 242011-7 Duplex Test Lead with Helical Spring-Point Hand Spikes
Representative image only.
Megger Mfr # 242011-7 – Item # EW-20011-04
Optimize performance of a multitude of Megger meters
  • 1 year warranty
$1,260.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)84
  • Length (cm)200
  • DescriptionDuplex Test Lead with Helical Spring-Point Hand Spikes, 7 ft
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The Megger 242011 series duplex test lead with helical spring-point duplex hand spikes is for use with Megger 247100, 247101, DLRO10HD, DLRO10, and DLRO10X digital low-resistance ohmmeters. This duplex test lead measures both current and resistance when taking four-wire resistance measurements, which are more accurate than two-wire resistance measurements and allow low resistance values to be detected. The lead has a maximum test current input of 10 amp.
Available for sale in the United States only