Megger 510006 Piezo Verifier, generates 10, 20 and 30 kV
Representative image only.
Megger Mfr # 510006 – Item # EW-20022-69
Safely confirm operation of voltage and phasing testers up to 30 kV
  • Small compact design easily allows the checking of the integrity of current paths and operation of indicators
  • Safely ensure IEC conformance
PRODUCT OPTIONS
$660.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • DC Voltage Range (Voltage)30000
  • DescriptionPiezo Verifier, generates 10, 20 and 30 kV
Includes
Lead and case.
Key Features
  • Small compact design easily allows the checking of the integrity of current paths and operation of indicators
  • Safely ensure IEC conformance
More About this Item
This essential safety tool is used to confirm the correct operation of voltage detectors and phasing testers that do not have a self-testing capability. When the verifier handle is depressed at the rate of 3 strokes per second, a piezo crystal generates a safety check voltage. This voltage, when applied to the tip of the detector, tests for integrity of the current path and proper operation of the indicators.
A three step proedure of testing the detector with the piezo verifier, using the detector to test for the absence of high voltage and then testing the detector again should be strictly observed in accordance with accepted operating procedures. Verifier can be used with single and two pole models.
Live chat
Alternatives
Alternatives