Megger BITE2 Battery Impedance Tester
Representative image only.
Megger Mfr # P30044-300 – Item # EW-20026-80
Advanced feature sets that include pass/warning/fail calculations based on a user-entered baseline value, advanced printing functions, and more
  • Determines condition of lead-acid and NiCd cells up to 7000 Ah
  • On-board Pass/Warning/Fail indications
  • Robust, repeatable instruments
  • On-line testing
  • Checks charger condition by measuring AC ripple current
  • Includes PowerDB LITE Software
PRODUCT OPTIONS
$6,964.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • DC Min (Volts)1
  • DC Max (Volts)25
  • DC Voltage Accuracy±(1.5% of rdg +1 LSD)
  • DisplayLCD, 2 x 16 characters
  • Safety RatingConforms to the EMC Directive 2004/108/EC and the LVD Directive 2006/95/EC
  • DescriptionBattery Impedance Tester
Key Features
  • Determines condition of lead-acid and NiCd cells up to 7000 Ah
  • On-board Pass/Warning/Fail indications
  • Robust, repeatable instruments
  • On-line testing
  • Checks charger condition by measuring AC ripple current
  • Includes PowerDB LITE Software
More About this Item
The BITE2 and BITE2P battery impedance testers determine the condition of lead-acid and nickel-cadmium cells up to 7000 Ah. The instruments work by applying a test current across the battery string while on-line, then measuring the total current (AC ripple and test current) and the voltage drop of each cell/jar. It then calculates the impedance. They also measure DC voltage and interconnection (strap) resistance to help determine the overall condition of the entire battery string's electrical path from terminal plate to terminal plate. The BITE2 and BITE2P receiver stores the readings in its internal memory. These measurements, along with other maintenance data such as ambient and pilot cell temperatures and AC ripple current, assist in determining the overall condition of battery systems.
Unlike load cycle testing that involves substantial downtime and repeated discharges, using these instruments requires no battery discharge, nor does it stress the battery in any way compared to other techniques. With a test time of less than 20 seconds for each cell and intercell connector, one person can easily, quickly, and precisely measure internal cell impedance, DC terminal voltage, and intercell connection resistance without taking the battery system off-line.