Megger CFL535G Dual-Channel Advanced Time Domain Reflectometer

Representative image only.
Megger Mfr # 1007-069 – Item # EW-20026-77
Get fast and accurate results when testing the integrity of communication, power, or control cables
  • Ensure the most effective parameters are selected with AUTO ranging option
  • Dual-channel, dual-trace, and dual-cursor capabilities provide instant distance between points
  • Use trace comparison to closely examine conditions between different traces
  • Store up to 100 trace measurements
  • Included TraceXpert software assists with downloading, reporting, and uploading
  • Can be used on energized circuits up to 150 V CAT IV or 300 V CAT III
  • 2 year warranty

$4,640.00USD / Each
sign in for your price.

SPECS & DESCRIPTION

Specifications & Description

  • Range10, 25, 50, 100, 250, 500, 1000, 2500, 5000, 10, , 20,000 m (30, 80, 160, 320, 800, 1600, 3200, 8000, 16,000, 32,500, 65,000 ft)
  • Accuracy±1% of range ±1 pixel at 0.67 VF
  • FunctionAUTO range, Dual trace, Dual cursor, Trace compare, Trace storage
  • Resolution1% of range
  • DisplayWVGA color graphics LCD (800 x 480 pixels), user-selectable color schemes; viewable in external environments
  • Battery Life12 hours (typical)
  • BatteryRechargeable lithium-ion (included)
  • Length (in)11 3/8
  • Width (in)7 1/2
  • Height (in)2 1/4
  • Power110 VAC for recharging batteries
  • DescriptionDual-Channel Advanced Time Domain Reflectometer
  • Warranty2 years
IP54 RatedTwo year warranty

Includes

Bed of nails test leads (2 pairs), TraceXpert software with USB cable, carrying case, Li-ion rechargeable battery, and AC/CD power supply and charger.

Key Features

  • Ensure the most effective parameters are selected with AUTO ranging option
  • Dual-channel, dual-trace, and dual-cursor capabilities provide instant distance between points
  • Use trace comparison to closely examine conditions between different traces
  • Store up to 100 trace measurements
  • Included TraceXpert software assists with downloading, reporting, and uploading
  • Can be used on energized circuits up to 150 V CAT IV or 300 V CAT III
  • 2 year warranty

More About this Item

These advanced dual-channel TDRs help locate cable faults for responsive or routine maintenance; quality control checks in manufacturing, shipping, and inventory management; and performance checks on cables in hard-to-reach or hazardous locations. The Auto option aids in rapid diagnosis of the TDR trace; manual override allows fine tuning and identification of difficult to determine faults.

Easily set up and view traces on a large, high-resolution color WVGA display. Directional control buttons and soft keys make it easy to use. Extra high resolution, backlight, and user definable tones and color provide vibrance to the graphical display, making it easier to identify events on the trace. Use trace comparison feature to closely examine conditions between different traces.

The TDRs store up to 100 traces in the internal memory. Recall the stored traces to the display for analysis or compare with an active display to help in fault location. Use the included TraceXpert software to download to a PC via the USB port for reporting and analysis. An IP54-rated case protects meter from dust and rain.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat