Megger DLRO10X Digital Low Resistance Microhmmeter; 10 Amp

Representative image only.
Megger Mfr # 1006-600; plus DH4-C – Item # EW-20022-30
Results in under 3 seconds with 0.2% basic accuracy
  • Water resistant to IP65
  • Auto current reversal cancels standing emfs
  • Fuse protected to 600V with 250 mW power limit (with optional override) to avoid heating the test sample
  • Automatically detects continuity in potential and current connections
  • Provides visible warning of voltages present at the terminals
  • Multiple operating modes including fully automatic
  • Menu system controlled by a two-axis paddle allows manual selection of maximum test current
  • Easy-to-read backlit LCD display
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • AC Current Range (mA)±10%
  • Max Resistance (ohms)1999.9
  • Resistance Resolution0.1 μΩ;1 μΩ;10 μΩ;100 μΩ;1 mΩ;10 mΩ;100 mΩ
  • Humidity90% RH @ 40ºC (104ºF) non-condensing
  • Min Temperature (° F)41
  • Max Temperature (° F)113
  • Min Temperature (° C)5
  • Max Temperature (° C)45
  • Data Logging700 tests
  • DisplayLCD
  • ComplianceIEC61010-1 600V Category III - only when DH6 leads are used
  • Battery Type (VAC)Rechargeable NiMH battery
  • Length (in)4
  • Height (in)9 1/2
  • DescriptionDatalogging Digital 10 Amp Low Resistance Ohmmeter
  • Warranty1 Year
1 Year WarrantyIP 65

Includes

NiMH battery pack and external charger.

Key Features

  • Water resistant to IP65
  • Auto current reversal cancels standing emfs
  • Fuse protected to 600V with 250 mW power limit (with optional override) to avoid heating the test sample
  • Automatically detects continuity in potential and current connections
  • Provides visible warning of voltages present at the terminals
  • Multiple operating modes including fully automatic
  • Menu system controlled by a two-axis paddle allows manual selection of maximum test current
  • Easy-to-read backlit LCD display
  • 1 year warranty

More About this Item

These digital low resistance microhmmeters provide accurate results in under three seconds. Select suitable test current, up to 10 A dc to measure reistance from 0.1 µΩ to 2000 Ω on one of sevent ranges. Consturcted with a strong, lightweight case, the units are well suited for use in the field or in the laboratory.

The DLRO10X unit features a RS-232 connector for downloading results in real time or to be stored for later retrieval. You can set high and low pass limints, enabling simple go-no-go testing.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat