Megger DLRO10X Digital Micro-Ohm Meter with Lead Set

Representative image only.
Megger Mfr # 1006-600 – Item # UX-20027-19
Accurate results in under three seconds
  • Auto current reversal cancels standing emfs
  • Fuse protected to 600 V
  • NiMH battery reduces weight
  • 250 mW power limit (with optional override) to avoid heating the test sample
  • Automatically detects continuity in potential and current connections
  • Visible warning of high voltages present at the terminals
  • Visible warning of current flowing in the test sample
  • Multiple operating modes including fully automatic
  • Alpha-numeric keypad for entering test notes
  • Users-selectable high and low limits
  • Auto-ranging with manual option
  • Printer output and memory

$5,625.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • DC Current Accuracy±10%
  • AC Current Accuracy±10%
  • Resistance Max (Ω)0.1
  • DisplayLarge color segment LED display
  • Memory700 tests
  • InterfaceReal time or from storage via RS-232
  • MeasurementManual, Auto, Continuous, Inductive, Undirectional
  • Safety RatingIn accordance with IEC61010-1, CAT III 600 V - only when DH6 leads are used
  • Width (in)3.9
  • Length (in)8.7
  • Height (in)9.3
  • Width (cm)10
  • Length (cm)22
  • Height (cm)23.7
  • WarrantyManufacturer Defect

Key Features

  • Auto current reversal cancels standing emfs
  • Fuse protected to 600 V
  • NiMH battery reduces weight
  • 250 mW power limit (with optional override) to avoid heating the test sample
  • Automatically detects continuity in potential and current connections
  • Visible warning of high voltages present at the terminals
  • Visible warning of current flowing in the test sample
  • Multiple operating modes including fully automatic
  • Alpha-numeric keypad for entering test notes
  • Users-selectable high and low limits
  • Auto-ranging with manual option
  • Printer output and memory

More About this Item

The DLRO10X digital micro-ohmmeter is a fully automatic instrument, selecting the most suitable test current, up to 10 A DC, to measure resistance from 0.1 µO to 2000 O on one of seven ranges. For users who desire more control over the measurement process, the DLRO10X uses a menu system controlled by a two-axis paddle to allow manual selection of the test current. It also enables the user to download results in real-time and save them to its on-board storage system for later download to a PC.

The strong, lightweight case is equally suitable for use in the field or in the laboratory. Light enough to be worn around the neck, it is small enough to be taken into areas which were previously too difficult to access.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat