Megger KC-C Low-Current Long Test Lead Set
Representative image only.
Megger Mfr # 1013-211 – Item # EW-20027-23
Easily and reliably measures lightning protection circuit resistance of tall structures
  • Designed to work with low-resistance ohmmeter 20027-21
  • Smaller and lightweight test leads provide safer working conditions
  • IP68-rated when test leads and terminations are locked together
  • Great connection flexibility—fully interchangeable instrument connections and test terminations
  • 1 year warranty
$1,445.00USD / Each
sign in for your price.
Often bought with...
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithMegger Ducter™ 2 A Low Resistance Ohmmeter 20027-21
  • DescriptionKC-C Low-Current Long Test Lead Set
  • Warranty1 year
1 Year Warranty
Key Features
  • Designed to work with low-resistance ohmmeter 20027-21
  • Smaller and lightweight test leads provide safer working conditions
  • IP68-rated when test leads and terminations are locked together
  • Great connection flexibility—fully interchangeable instrument connections and test terminations
  • 1 year warranty
More About this Item
This test lead set includes a cable reel that extends 164 ft long—ideal for measuring the circuit resistance of tall and long structures. Use the test leads with low-resistance ohmmeter 20027-21 to test the lightning protection of a wind turbine and lightning and static protection on airplanes. With the low current capability, the test lead cables can be thinner, allowing the cables to be smaller and lighter, helping improve worker safety when working from a great height.
The test lead set consists of two cable leads, a 9.8-ft long cable and a 164-ft long cable on a reel. All instrument connections are interchangeable with the test connections, which allow for great flexibility when connecting to a wind turbine for measurements. The connections are IP68-rated once click locked in place; simply align and push together to make the waterproof seal.